Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ DAPI Nuclear Counterstain
Highly pure diamidino-2-phenylindole (DAPI) dye for fluorescent staining of DNA content and nuclei in fixed-cell imaging techniques.
$208.80 - $268.90
Specifications
Detection Fluorescent
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
PI62247
thermo scientific™
62247
10mg Each for $208.80
PIEN62248
thermo scientific™
62248*
1mL Each for $268.90
Description & Specifications
Specifications
Detection Fluorescent
Thermo Scientific™ Pierce DAPI Nuclear Counterstains are high-purity forms of diamidino-2-phenylindole dye for fixed-cell, fluorescent staining of DNA content and nuclei for cellular imaging techniques.
As a counterstain in fluorescence imaging methods, DAPI is compatible with antibodies and other probes labeled with fluorescein and rhodamine dyes, as well as with Thermo Scientific DyLight™ Fluors. DAPI has greater photostability than Hoechst dyes, although Hoechst 33342 can be use for live cell imaging while use of DAPI is confined to fixed cells. DAPI is offered in powdered solid and aqueous solution forms.
Highlights:
  • Nuclei detection– binds specifically and quantitatively with DNA to facilitate specific detection of nuclei and DNA content in fluorescence imaging
  • Fluorescent– yields blue fluorescent product to enable measurement with fluorescence microscopes and imagers (excitation 341nm, emission 452nm)
  • Fixed cells– especially for use with fixed cells rather than live-cell samples
  • Compatible counterstain–provides effect counterstaining alongside fluorescein and other fluorescent probes, such as Thermo Scientific DyLight Fluors
Recommended for:
Assaying DNA in solution; Diagnosing mycoplasmal infection of cell cultures; Measuring nuclear content and sorting chromosomes in flow cytometry; Assessing apoptosis; Detecting nuclei and organellar DNA in immunofluorescent and in situ hybridization procedures; Replacing ethidium bromide for staining DNA in agarose gel; Counterstaining nuclei in histochemical methods when red-fluorescent antibodies have been used to detect specific targets; Reports also indicate that DAPI will bind to polyphosphates and other polyanions, dextran sulfate and SDS.