Thermo Scientific™ Pierce™ Coomassie Plus™ (Bradford) Protein Assay
Brown-to-purple (A595nm), ready-to-use, fast reducing agent-compatible assay reagent to measure total protein concentration vs. protein standard.
$236.95 - $296.48
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI23236 Thermo Scientific™
PI23236
Pierce Coomassie Plus (Bradford) Protein Assay Kit Pack for $296.48
PI23238 Thermo Scientific™
PI23238
Pierce Coomassie Plus (Bradford) Protein Assay Reagent Each for $236.95
Specifications
Description
Thermo Scientific™ Pierce Coomassie Plus Protein Assay is a high-performance Bradford assay reagent — one of the fastest, simplest and most popular protein quantitation methods available.
The Pierce Coomassie Plus Assay Reagent is a single, ready-to-use solution for measuring protein concentration. Simply add the reagent to equal volumes of samples and standards, mix, then measure the absorbance at 595nm. The assay costs only pennies per sample and can be performed in either test tube or microplate format. The Pierce Coomassie Plus Assay Reagent provides increased linearity of response and only half the protein-to-protein variability of other commercial Bradford assay formulations.
Highlights:
  • Colorimetric – measure with a standard spectrophotometer or plate reader (595nm)
  • Easy to use – single reagent; no working reagent preparation required
  • Fast – almost immediate color development; add, mix, and read results
  • Broad range – detects protein concentration in the range 1 to 1500μg/mL
  • Better – improved linearity and response-uniformity compared to traditional Bradford formulations
  • Flexible – microplate and cuvet protocols provided with the instructions and adaptable to several target working ranges
Includes:
Kit contains Protein Assay Reagent and 10 ampules of Albumin Standard