Thermo Scientific™ Pierce™ Compat-Able™ Protein Assay Kits
Precipitation reagents to eliminate interfering substances from protein assay samples to make them compatible with BCA and Coomassie-Bradford assays.
$188.67 - $445.10
Specifications
Kit Contents Compat-Able Reagent 1, 250mL,Compat-Able Reagent 1, 250mL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI23215 Thermo Scientific™
PI23215
Compat-Able Protein Assay Preparation Reagent Set Each for $188.67
PI23229 Thermo Scientific™
PI23229
Compat-Able BCA Protein Assay Kit Each for $344.20
PI23239 Thermo Scientific™
PI23239
Compat-Able Coomassie Plus Protein Assay Kit Each for $445.10
Specifications
Description
Thermo Scientific™ Compat-Able Protein Assay Preparation Set eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.
The Compat-Able Reagents remove salts, detergents, reducing agents and other substances from protein samples to eliminate interference with protein assays. The protocol selectively precipitates all of the protein, thereby allowing the non-protein components to be easily decanted and removed. The purified, precipitated protein is redissolved in water, making it ready for accurate determination of the protein concentration by standard protein assay methods. The procedure is accomplished in less than 10 minutes with just a few simple steps. The Compat-Able Preparation Set is available individually or conveniently bundled with either the Thermo Scientific Pierce BCA Protein Assay Kit or the Thermo Scientific Pierce Coomassie Plus Protein Assay Kit.
Highlights:
  • Eliminate incompatibilities – removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer
  • Simple and fast – prepare samples in less than 10 minutes with four simple steps
  • Convenient – reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results
  • Scalable – microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs
  • Validated – tested and offered together with BCA and Coomassie Plus Protein Assay Kits; protocol integrates control and normalization procedures
Includes:
Set contains two bottled reagents (Reagent 1 and Reagent 2); Complete kits contain the Reagent Set plus components of the respective protein assay
Description & Specifications
Specifications
Compat-Able Reagent 1, 250mL,Compat-Able Reagent 1, 250mL