missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Coated Plate IP Kits

Perform immunoprecipitation assays without resins, beads, centrifugation or magnets using coated streptavidin or Protein A/G 96-well microplates.

$567.32 - $578.52

Specifications

Quantity 2 plate kit
Formulation Multi-component kit
Kit Contents Neutralization Buffer, 12mL,Neutralization Buffer, 12mL,Neutralization Buffer, 12mL,Neutralization Buffer, 12mL,Neutralization Buffer, 12mL
Sufficient For Assaying 48 samples with controls or 192 total IP reactions
Support Type Polystyrene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI45350 Thermo Scientific™
PI45350
Pierce Protein A/G Coated Plate IP Kit Each for $567.32
PI45360 Thermo Scientific™
PI45360
Pierce Streptavidin Coated Plate IP Kit Each for $578.52
Specifications

Description

Thermo Scientific™ Pierce Coated Plate IP Kits enable immunoprecipitation experiments to be performed in 96-well microplates.

These coated microplate immuunoprecipiation kits provide for rapid immunoprecipitation of multiple samples without the usual tedium of pipetting, centrifuging and separating beaded affinity resin in individual microcentrifuge tubes. Immunoprecipitation is accomplished using coated 96-well microplates rather than beaded agarose resin. The plate format allows for faster processing of multiple samples. The Protein A/G Coated Plate Kit can be used for immunoprecipation with any antibody that binds to Protein A or G. The Streptavidin Coated Plate Kit is for immunoprecipitation with biotinylated IP antibodies.

Highlights:

  • Ready-to-use, high quality coated plates provide high capacity and consistency
  • Accommodate antibody and sample volumes comparable to tube-based methods
  • Easily recover immunoprecipitated samples for downstream analysis
  • Coated plates are 96-well strip plates, convenient for experiments requiring only a partial plate
  • Include simple instructions, including detailed explanation of appropriate controls
  • Two kits available, suitable for most common antibody types (mouse, rabbit, human and goat IgG subclasses) or any biotinylated antibody or bait protein

Advantages of the Plate Format:

  • Best suited for simultaneously processing multiple samples and their control conditions
  • Allows faster, easier and more thorough washing than with traditional tube/resin IP methods
  • Makes use of familiar and convenient ELISA tools (multichannel pipettors and plate washing)
  • No no tubes to open and close, and no centrifugation required

Protein A/G Kit (Part No. 45350)

  • Binds all human IgG subclasses; human IgM, IgA (weak), and IgE; and all mouse IgG subclasses
  • Offers increased binding for mouse monoclonal antibodies compared to Protein A or Protein G alone
  • Free of pH dependency when binding IgGs, providing more flexible binding conditions

Streptavidin Kit (Part No. 45360)

  • Used in combination with biotinylated antibodies, resulting in IP and co-IP preparations free of interfering antibody fragments
  • Contains biotin-blocking buffer to minimize endogenous biotin binding

Includes:

Coated 96-well strip plates, uncoated 96-well strip plates, PBS pack, detergent solution, elution buffer, neutralization buffer and sealing tape

Recommended for:

Rapid, trouble-free immunoprecipitation of multiple samples and their controls; Immunoprecipitation when plate-based tools and techniques are preferred

Description & Specifications

Specifications

2 plate kit
Neutralization Buffer, 12mL,Neutralization Buffer, 12mL,Neutralization Buffer, 12mL,Neutralization Buffer, 12mL,Neutralization Buffer, 12mL
Polystyrene
Multi-component kit
Assaying 48 samples with controls or 192 total IP reactions