Thermo Scientific™ Pierce™ C-18 Tips
Capture, concentrate and desalt 10 to 100 microliter samples of peptides and proteins for MALDI mass spectrometry with these ready-to-use pipette-tip columns of C18 resin.
$63.92 - $407.43
Specifications
Hazards Irritant
Standard Components Silica
Standard Concentration 98 to 100%
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Quantity Price Quantity  
Catalog Number Mfr. No. Description Quantity Price Quantity  
PI87781 Thermo Scientific™
87781
Pierce C-18 Tips, 10uL bed 8 tips Pack of 8 Each for $63.92
PI87782 Thermo Scientific™
87782
Pierce C-18 Tips, 10uL bed 96 tips Pack of 96 Each for $358.55
PI87784 Thermo Scientific™
87784
Pierce C-18 Tips, 100uL bed 96 tips Pack of 96 Each for $407.43
Specifications
Description
Thermo Scientific™ Pierce C-18 Tips are ready-to-use pipette-tip columns of C18 resin that enable fast and efficient capture, concentration, desalting and elution of peptides and proteins for MALDI mass spectrometry and other methods.
Improve protein analysis results with Pierce C18 Tips by removing urea, salts and other contaminants before MS analysis. The tips are ideal for preparing samples for matrix-assisted laser desorption ionization (MALDI) or nanoelectrospray ionization techniques. The Pierce C18 Tips are available in convenient 10 and 100μL tips with binding capacities of 8 and 80μg, respectively.
Highlights:
  • Monolithic C18 fast-flow tips have low resistance and improved flow characteristics compared to other commercially available tips
  • High binding capacity in a convenient tip format
  • Two pipette tip formats for processing up to 8 or 80μg of sample
  • Eight packs and 96-tip trays are automation compatible
Description & Specifications
Specifications
Irritant
Silica
98 to 100%