Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ C-18 Tips


Capture, concentrate and desalt 10 to 100 microliter samples of peptides and proteins for MALDI mass spectrometry with these ready-to-use pipette-tip columns of C18 resin.

Manufacturer: thermo scientific™  87781

 View more versions of this product

Catalog No. PI87781

  • / Pack of 8 Each
 

Description & Specifications

Specifications

Storage Room Temperature
Hazards Irritant
Standard Components Silica
Quantity 8 tips
Standard Concentration 98 to 100%
Storage Requirements Room Temperature
Sufficient For Processing 8 samples (10µL each) containing up to 8µg peptide each
Description Pierce C-18 Tips, 10uL bed
Formulation Pipette tips containing 10µL of porous C-18 reversed-phase resin
For Use With (Application) MALDI-MS
For Use With MALDI-MS

Thermo Scientific™ Pierce C-18 Tips are ready-to-use pipette-tip columns of C18 resin that enable fast and efficient capture, concentration, desalting and elution of peptides and proteins for MALDI mass spectrometry and other methods.

Improve protein analysis results with Pierce C18 Tips by removing urea, salts and other contaminants before MS analysis. The tips are ideal for preparing samples for matrix-assisted laser desorption ionization (MALDI) or nanoelectrospray ionization techniques. The Pierce C18 Tips are available in convenient 10 and 100μL tips with binding capacities of 8 and 80μg, respectively.

Highlights:

  • Monolithic C18 fast-flow tips have low resistance and improved flow characteristics compared to other commercially available tips
  • High binding capacity in a convenient tip format
  • Two pipette tip formats for processing up to 8 or 80μg of sample
  • Eight packs and 96-tip trays are automation compatible