Thermo Scientific™ Pierce™ BS2G-d0 and BS2G-d4 Crosslinker Mass Pair
Crosslink amines to amines using these heavy (deuterated) and light matched pairs of a 7.7A, NHS-ester crosslinker for mass spectrometry analysis of protein interactions.
$257.04 - $556.50
Specifications
Quantity 10mg
Sufficient For 19mL of typical 1X (1mM) reaction solution, or approx. 5 uses at 2mg reagent per use
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI21610 Thermo Scientific™
PI21610*
Pierce BS2G-d0 Each for $257.04
PI21615 Thermo Scientific™
PI21615*
Pierce BS2G-d4 Each for $556.50
Specifications
Description
Thermo Scientific™ Pierce BS2G-d0/d4 reagents form a matched set of heavy (deuterated) and light mass varieties of a short, amine-reactive crosslinker that facilitates the identification of crosslinked peptides by mass spectrometry.
Highlights:
  • Well-characterized, high-purity, deuterium-labeled crosslinker and its hydrogen-containing analog
  • Instructions edited by an expert in the crosslinking/MS strategy field
  • Suberate- and glutarate-based reagent pair - provides a “molecular ruler” option to study inter- and intra-molecular distances to gain structural information not possible with just one reagent
  • Efficient and convenient - requires only microgram amounts of protein; an excellent alternative to NMR or X-ray crystallography-based methods that require large amounts of protein, special solvents or crystal formation
  • Reactive groups: Sulfo-NHS ester (homobifunctional)
  • Reactive toward: amino groups
Description & Specifications
Specifications
10mg
19mL of typical 1X (1mM) reaction solution, or approx. 5 uses at 2mg reagent per use