Thermo Scientific™ Pierce™ Bovine Serum Albumin Standards
High-quality, calibrated BSA solutions (2mg/mL) to serially dilute to create protein assay standard curves to accurately measure protein concentration.
$104.46 - $216.56
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Quantity Price Quantity  
Catalog Number Mfr. No. Description Quantity Price Quantity  
PI23208 Thermo Scientific™
PI23208
Pierce Bovine Serum Albumin Standard Pre-Diluted Set 7 x 3.5mL Each for $216.56
PI23209 Thermo Scientific™
PI23209
Pierce Bovine Serum Albumin Standard Ampules, 2mg/mL 10 x 1mL Pack of 10 Each for $104.46
PI23210 Thermo Scientific™
PI23210
Pierce Bovine Serum Albumin Standard, 2mg/mL 50mL Pack for $106.43
Specifications
Description
Thermo Scientific™ Pierce BSA Protein Assay Standards are high-quality reference samples for generating accurate standard curves and calibration controls in total protein assays.
These bovine serum albumin (BSA) solutions are protein concentration reference standards for use in BCA, Bradford and other protein assay protocols. BSA is the universally accepted reference protein for total protein quantitation. The albumin standard is precisely formulated at 2mg/mL in an ultrapure 0.9% sodium chloride (saline) solution. The concentration of the stock solution is calibrated by direct comparison to purified BSA (Fraction V) from the National Institute of Standards and Technology (NIST™). The product is available in three convenient package formats: 1mL glass ampules, 50mL polypropylene bottles, and complete sets containing seven ready-to-use dilutions.
Highlights:
  • Universal – recognized as the industry standard for protein quantitation in colorimetric protein assays
  • Pure and stable – supplied in ultrapure 0.9% saline solution with 0.05% sodium azide; room temperature stable
  • Accurate and consistent – precisely formulated at 2.00 +/-0.03mg/mL compared to a NIST reference
  • Convenient – choose 1mL ampules, a 50mL bottle, or a complete set containing seven ready-to-use dilutions
Includes:
2mg/mL BSA solution (Part No. 23209, 23210) or 7-solution set of different concentrations (Part No. 23208)
Recommended for:
Protein assay quantitation standard; Protein recovery control for desalting and other column procedures; General calibration of spectrophotometer UV-lamp (A280)