Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Amino Acids for SILAC Labeling


Study protein expression by mass spectrometry with mass variants of lysine, arginine and other amino acids to perform stable isotope labeling with amino acids in cell culture.

Manufacturer: thermo scientific™  88436

 View more versions of this product

Catalog No. PI88436

  • / Each
 

Description & Specifications

Specifications

Quantity 500mg
Formulation Lyophilized powder of L-Leucine with at least 99% 13C6
Sufficient For Media for 10 SILAC experiments
Description L-Leucine, 13C6
For Use With (Application) SILAC experiments (stable isotope labeling with amino acids in cell culture)

Thermo Scientific™ Heavy and Light Amino Acids are used to specifically analyze protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC) quantification kits.

SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of “heavy“ 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.

Highlights:

  • Efficient – 100% label incorporation into proteins of living cells

  • Flexible – different isotopes of heavy and/or light amino acids for arginine, lysine, leucine and proline enable the quantitation of peptides derived from MS-grade proteases

  • Multiplex capabilities – several alternative isotopes of arginine and lysine are available that allow the analysis of multiple treatment conditions in each experiment

  • High-quality supplements – heavy amino acids with >99% isotope purity