Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ AMCA Fluorescent Dyes


Label antibodies, proteins and other molecules for fluorescence imaging with these activated forms of aminomethyl coumarin acetate (AMCA) blue-fluorescent dye.

Manufacturer: thermo scientific™  PI33015

 View more versions of this product

Catalog No. PI33015

  • / Pack
 

Description & Specifications

Specifications

Description Pierce Hydrazide-AMCA
Quantity 5mg
Sufficient For 1 or 2 uses to label a total of 101mg of IgG using typical conditions
Formulation Hydrazide-activated aminomethylcoumarin acetate

Thermo Scientific™ AMCA Reagents are derivatives of AMCA blue fluorescent dye that are activated for easy and reliable labeling of antibodies, proteins and other molecules for use as fluorescent probes.

AMCA (aminomethylcoumarin acetate) is a blue fluorescent dye with reactive derivatives that are used as contrasting probes for double- and triple-labeling in immunofluorescence microscopy, arrays and in situ hybridization. The desirable properties of AMCA dyes include a relatively large Stoke's shift and resistance to photobleaching. NHS-AMCA and Sulfo-NHS-AMCA are reactive towards primary amine groups on proteins, peptides and other biomolecules. AMCA-Hydrazide is used to label glycosylation sites or for conjugation to carboxyl groups using the crosslinker EDC. AMCA has an excitation maximum at 345nm and an emission maximum at 450nm.

Highlights:

 AMCA — activated derivatives of blue-fluorescent aminomethylcoumarin acetate dye

Amine-specific labeling — NHS-ester and Sulfo-NHS-ester varieties of AMCA efficiently label antibodies and other purified proteins at primary amines (lysine side chains)

Glycoprotein labeling — hydrazide variety of AMCA labels oxidized sugar groups in glycoproteins or other carbohydrates