Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ Active GTPase Pull-Down and Detection Kits
Purify GTPases and assay activation levels using specific GST-tagged protein-binding domains (PBD), GSH-agarose beads and anti-GTPase antibodies.
$1,201.51 - $1,202.46
Specifications
Quantity 30 test kit
Sufficient For 30 pull-down assays from 0.5 to 1mg cell lysate samples
View More Specs
 Disclaimers
Products
Catalog Number Mfr. No. Description Kit Contents Price Quantity    
PI16116
thermo scientific™
16116
Pierce Active Rho Pull-Down and Detection Kit Anti-Rho Antibody, 5 units, 75uL Each for $1,201.51
PI16117
thermo scientific™
16117
Pierce Active Ras Pull-Down and Detection Kit Anti-Ras Antibody, 5 units, 250µL Each for $1,202.46
PI16118
thermo scientific™
16118
Pierce Active Rac1 Pull-Down and Detection Kit Anti-Rac1 Antibody, 5 units, 50µL Each for $1,201.51
PI16119
thermo scientific™
16119
Pierce Active Cdc42 Pull-Down and Detection Kit Anti-Cdc42 Antibody, 5 units, 300µL Each for $1,201.51
PI16120
thermo scientific™
16120
Pierce Active Rap1 Pull-Down and Detection Kit Anti-Rap1 Antibody, 5 units, 50µL Each for $1,201.51
PI16121
thermo scientific™
16121*
Pierce Active Arf1 Pull-Down and Detection Kit Anti-Arf1 Antibody, 5 units, 25µL Each for $1,201.51
Description & Specifications
Specifications
Quantity 30 test kit
Sufficient For 30 pull-down assays from 0.5 to 1mg cell lysate samples
Measure activation of small GTPases by isolating them via their specific downstream effectors with Thermo Scientific™ Pierce Active GTPase Pull-Down and Detection Kits.
The Active GTPase Pull-Down and Detection Kits measure activation of small GTPases by isolating them via their specific downstream effectors. Kits specific for Arf1, Arf6, Cdc42, Rac1, RalA, Rap1, Ras, and Rho GTPases are offered. The respective binding domain of the downstream effector for each small GTPase is expressed as a GST-fusion protein. When immobilized on a resin, this fusion protein is used to pull down the active (i.e., GTP-bound) GTPase. The pull-down affinity purification step is easily and conveniently performed using microcentrifuge spin columns. After a simple wash step, the bound GTPase is recovered by eluting the GST-fusion protein from the glutathione resin using SDS-PAGE sample loading buffer. The purified GTPase is then detected by Western blot using a specific antibody supplied in each kit.
Each GTPase Kit includes GST-fusion protein of the specific binding domain, GTPγS and GDP activators, glutathione agarose resin, lysis-binding-wash buffer, primary antibody, 2X SDS sample buffer, spin columns and collection tubes. Each kit contains sufficient components for 30 pull-down assays. The kits are optimized for Western blot detection with an HRP-conjugated secondary antibody (Part No. 31460, rabbit; Part No. 31430, mouse) using Thermo Scientific SuperSignal™ West Pico Chemiluminescent Substrate (Part No. 34080).
Highlights:
  • High sensitivity and accuracy – optimized reagents, specific antibodies and Western blot procedure ensure accurate controls and quantitative results
  • Convenience – no need to express and purify your own GST-PBD or RBD fusion proteins or use expensive antibody-based immunoprecipitation affinity resins
  • Speed – simultaneous incubation of cell lysate with GST-PBD or RBD and the glutathione resin directly in a high-performance microcentrifuge spin column
  • Ease-of-use – reagents and pull-down conditions have been optimized for immediate success, even for first-time users
  • Efficiency – the microcentrifuge spin columns and collection tubes provide efficient separation of liquid and resin, preventing sample loss
  • Complete – kits include reagents for affinity purification as well as controls, a compatible cell lysis buffer and specific primary antibody for final Western blot detection
GST fusion proteins of active GTPase binding domains:
  • Raf1 Ras-binding domain (Raf1-RBD) binds active Ras
  • Pak1 p21-binding domain (Pak1-PBD) binds active Rac1 and Cdc42
  • Rhotekin Rho-binding domain (Rhotekin-RBD) binds active Rho
  • RalGDS Rap1-binding domain (RalGDS-RBD) binds active Rap1
  • GGA3 protein-binding domain (GGA3-PBD) binds active Arf1 and Arf6
  • Ral binding protein 1 (RalBP1) binds active RalA
Includes:
GST fusion protein of specific binding domain, glutathione agarose resin, GTPγS and GDP (100X), lysis-binding-wash buffer, GTPase-specific primary antibody, SDS-PAGE sample loading buffer (2X), spin columns and collection tubes