Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Acetonitrile (ACN), LC/MS Grade

Thermo Scientific™ Pierce Acetonitrile (ACN) is an LC/MS-grade preparation with high purity and low UV-absorptivity that makes it suitable for HPLC and mass spectrometry applications.

$588.04 - $588.04

Specifications

Grade LC/MS grade
Formulation Clear, colorless liquid of acetonitrile
For Use With HPLC/MS
Hazards Flammable, Irritant, Toxic
Standard Components Acetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

PI85188

 
thermo scientific™
85188
4 x 1L Pack of 4 Each for $588.04
Description & Specifications

Specifications

Grade LC/MS grade
Formulation Clear, colorless liquid of acetonitrile
For Use With HPLC/MS
Hazards Flammable, Irritant, Toxic
Standard Components Acetonitrile
Standard Concentration 98 to 100%
Storage Room Temperature

Thermo Scientific™ Pierce Acetonitrile (ACN) is an LC/MS-grade preparation with high purity and low UV-absorptivity that makes it suitable for HPLC and mass spectrometry applications.

Thermo Scientific Pierce LC/MS Grade Acetonitrile is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance to provide you with the most sensitive detection across all wavelengths. Pierce Acetonitrile is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmosphere with PTFE-lined fluorocarbon caps for ultimate protection.

Highlights:
  • 37 quality tests ensure low, stable baselines and lot-to-lot consistency
  • Low UV absorbance yields low baselines and high detection sensitivity
  • Low impurities protects columns and simplifies analysis by eliminating extraneous peaks

Product Details:

  • Alternative names: Methyl cyanide, cyanomethane, ethanenitrile
  • Molecular formula: CH3CN
  • Molecular weight: 41.05
  • Density: 0.780g/mL
  • CAS number: 75-05-8

Pierce LC/MS Grade Acetonitrile is specially purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and to prolong the life of your equipment. These Acetonitrile specifications also meet ACS standards.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.