Thermo Scientific™ Pierce™ ABC Staining Kits
Detect biotinylated secondary antibodies by IHC using the highly sensitive avidin-biotin complex (ABC) technique.
$456.75 - $841.00
Specifications
Detection Chromogenic
Kit Contents
Reagent A (Avidin), 2mL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI32020 Thermo Scientific™
PI32020
Pierce ABC Peroxidase Standard Staining Kit
Pack for $456.75
PI32050 Thermo Scientific™
PI32050
Pierce Ultrasensitive ABC Standard Peroxidase Staining Kit Pack for $588.70
PI32052 Thermo Scientific™
PI32052
Pierce Ultrasensitive ABC Peroxidase Mouse IgG Staining Kit Pack for $841.00
PI32054 Thermo Scientific™
PI32054
Pierce Ultrasensitive ABC Peroxidase Rabbit IgG Staining Kit Pack for $835.20
Specifications
Description
Thermo Scientific™ Pierce ABC Staining Kits include reagents for the avidin-biotin complex (ABC) technique, a highly sensitive method for immunohistochemical detection with biotinylated secondary antibodies.
Highly diluted primary antibodies can be used with ABC Staining Kits, producing comparable stain intensity to other methods that require higher concentrations of antibody. Kits with a secondary antibody are selected by the species of primary antibody to be used. For example, if the primary antibody (IgG) is produced in mice, select the ABC Mouse IgG Kit. All you need to create a sensitive detection system is your specific primary antibody, an ABC Kit with biotinylated antibody and a substrate for horseradish peroxidase.The Standard Kits include only the avidin and biotinylated enzyme. They are useful if an ABC Kit is not available for your specific species of primary antibody, or if the primary antibody is directly labeled with a Thermo Scientific EZ-Link Biotinylation Reagent. The biotinylated antibody for your specific application can then be used, along with blocking serum and a Standard ABC Kit.
Highlights:
  • Produce very low background staining
  • Require lower primary antibody concentrations than standard methods
  • Ideal for immunohistochemistry, ELISA, and Western blotting applications
ABC Kits
  • Available with horseradish peroxidase
  • Contain a biotinylated secondary antibody in a choice of species – the species should be selected based on the species of the primary antibody to be used
Standard Kits
  • Available with a choice of alkaline phosphatase or horseradish peroxidase
  • Suitable for use when an ABC kit is not available for the species of the primary antibody or if the specificity of the antibody in the kit is not appropriate for the application
  • Do not include secondary antibody or blocking serum – secondary antibody and blocking serum appropriate to the application should be purchased separately
Ultrasensitive ABC Kits
  • Available with horseradish peroxidase
  • Provide greater sensitivity than ABC Kits without increased background staining
  • Supply the extra sensitivity needed for localizing antigens present in very small quantities
  • Permit approximately five-fold higher dilutions of primary antibody to be used than do ABC Kits
Includes:
Biotinylated affinity-purified secondary antibody, 1mL (except Standard Kits); Appropriate blocking serum, 3mL (except Standard Kits); Avidin reagent, 2mL; Biotinylated enzyme reagent, 2mL
Description & Specifications
Specifications
Chromogenic
Reagent A (Avidin), 2mL