Thermo Scientific™ Pierce™ 96-Well Microplates
Bind antibodies with high capacity using these specially formulated polystyrene plates for ELISA and other immunoassays.
$181.50 - $576.06
Specifications
Sufficient For ELISA and other plate based assays requiring protein binding
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI15041 Thermo Scientific™
PI15041
Pierce 96-Well Polystyrene Plates, Corner Notch Pack of 100 Each for $576.06
PI15042 Thermo Scientific™
PI15042
Pierce 96-Well Polystyrene Plates, White Opaque Pack of 25 Each for $181.50
Specifications
Description
Thermo Scientific™ Pierce 96-Well Microplates are made of specially formulated polystyrene and treated to produce high-capacity antibody-binding characteristics.
Pierce 96-Well Microplates are standard flat-bottom clear and white assay plates for ELISA and other protein-based methods. Each lot is certified to ensure adherence to strict performance criteria. Clear and opaque white varieties are available in 100-plate and 25-plate cases, respectively. Use the clear plates to perform colorimetric ELISA and other assays for measurement in standard spectrophotometer plate readers. Use the white plates to perform fluorescent or chemilumiescent assays for detection in fluorescence or luminometer readers.
Highlights:
  • Specially formulated optically clear polystyrene and treated to provide protein coating capacity
  • High binding capacity surface improves the overall performance in immunoassay applications
  • Certified for performance, having consistently low coefficients of variation (CV) among wells
Description & Specifications
Specifications
ELISA and other plate based assays requiring protein binding