Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ 660nm Protein Assay
Red-to-green (A660nm), ready-to-use, detergent- and reducing agent-compatible assay reagent to measure total protein concentration vs. protein standard.
$151.90 - $325.88
Products
Catalog Number Mfr. No. Description Price Quantity    
PI22660
thermo scientific™
22660
Pierce 660nm Protein Assay Reagent Each for $244.45
PI22662
thermo scientific™
22662
Pierce 660nm Protein Assay Kit Each for $325.88
PI22663
thermo scientific™
22663
Pierce Ionic Detergent Compatibility Reagent Each for $151.90
Description & Specifications
Thermo Scientific™ Pierce 660nm Protein Assay is a quick, ready- to-use colorimetric method for total protein quantitation.
Additional Information and Online Ordering »
The Pierce 660nm Protein Assay is a quick, ready-to-use colorimetric method for measuring protein concentration. The assay is more linear than coomassie-based Bradford assays and compatible with higher concentrations of most detergents, reducing agents and other commonly used reagents. The accessory Ionic Detergent Compatibility Reagent (IDCR) provides for even broader detergent compatibility, making the Pierce 660nm Protein Assay the only protein assay that is suitable for samples containing Laemmli SDS sample buffer with bromophenol blue.
Although the Pierce 660nm Protein Assay produces a higher level of protein-to-protein variation (37%) than other assays, such as the BCA Protein Assay, the simpler single-reagent format and broader substance compatibility make the 660nm Assay more convenient for many routine applications. The Pierce 660nm Protein Assay can be performed in either a test tube or microplate format.
The assay is based on a proprietary dye-metal complex that binds to proteins in an acidic medium. Upon binding, the reddish dye-metal complex turns to green, resulting in an absorbance shift that is measurable at wavelengths near 660nm (645 to 670). The dye interacts mainly with basic residues in proteins, such as histidine, arginine and lysine and to a lesser extent tyrosine, tryptophan and phenylalanine.
Highlights:
  • Versatile – works with a greater range of detergents and reducing agents than other dye-based assays
  • Fast – single reagent with a simple mix-and-read protocol; no working reagent to prepare
  • Accurate – produces standard curves that are more linear than with the Bradford assay method
  • Flexible – assay may be performed in test tubes or microplates
  • Conserve samples – requires only 10μL for microplate or 100μL for the test tube procedures
  • Convenient – room temperature storage means no waiting for reagent equilibration before use
Includes:
Kit contains Pierce 660nm Assay Reagent and a set of seven pre-diluted albumin protein assay standards