Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ 10K MWCO 96-well Microdialysis Plate
Simultaneously process 1 to 96 samples with 10 to 100µL volumes with this convenient and disposable rapid dialysis system that is automation compatible.
$18.45 - $872.63
Products
Catalog Number Mfr. No. MWCO Sufficient For Quantity Price Quantity    
PI88260
thermo scientific™
88260
10000 Da Dialysis of 96 samples 10 to 100uL each 1 plate set Each for $872.63
PI88262
thermo scientific™
88262
3500 Da Dialysis of 96 samples 10 to 100uL each 1 plate set Each for $716.11
PI88269
thermo scientific™
88269
---- 12 x 8 microdialysis plate wells 1 sheet Each for $18.45
Description & Specifications
The Thermo Scientific™ Pierce 96-well Microdialysis Plate, 10K MWCO is a convenient and disposable rapid dialysis system for the simultaneous processing of 1 to 96 samples with volumes 10 to 100μL and is compatible with automation systems.
Each sample processed with the Pierce 96-well Microdialysis Dialysis Plate is dialyzed inside a single, removable dialysis device created by two 10K MWCO regenerated cellulose membranes separated by <2mm. The short diffusion distance and large surface area facilitates rapid dialysis with the removal of salts and small molecules often completed in 2 to 4 hours. Sample recovery is performed using standard laboratory micropipettes. Individual microdialysis devices can also be used with 2mL microcentrifuge tubes for processing single samples.
Highlights:
  • High yield – greater than 90% protein recovery after dialysis
  • Rapid – dialysis completed in 2 to 4 hours
  • Easy to use – sample loading and retrieval can be done with a standard pipette
  • Flexible – detachable 8-device strips, scalable from 1 to 96 samples
  • Automation-compatible – plate format conforms to the SBS Microplate Standard
Recommended for:
Protein and DNA desalting/buffer exchange; Removal of dyes after protein labeling; Enzyme activity/binding assays; Drug binding studies