Thermo Scientific™ Pierce™ 1.1mL Microtubes and Racked Systems
Pre-racked 1.1mL-tall microtubes boxed to match a standard 96-well microplate footprint for multichannel pipetting or automated workstations.
$67.31 - $161.63
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI15082 Thermo Scientific™
PI15082
Pierce 1.1mL Microtube Racked System Pack of 10 Each for $161.63
PI15086 Thermo Scientific™
PI15086
Pierce 1.1mL Microtubes Pack of 960 Each for $93.94
PI15088 Thermo Scientific™
PI15088
Pierce 1.1mL Microtube In-Line Caps Pack of 120 Each for $67.31
Specifications
Description
Thermo Scientific™ 1.1mL Microtubes and Racked Systems consist of 1.1mL tall microtubes supplied pre-racked in boxes to match a standard 96-well microplate footprint.
The microtubes resembles 1mL pipette tips except that they have round bottoms. Like a box of 1mL pipette tips, the racked microtubes match multi-channel pipettors, providing convenience for a variety of reagent- and sample-dispensing procedures.
Highlights:
  • 8 x 12 format simplifies and speeds applications in which a multi-channel pipettor is used
  • Format is also compatible with robotic workstations
  • Thermo Scientific Microtube System can be centrifuged