PIERCAN USA Polyurethane/Chlorosulfonated Polyethylene Long Gloves DFS Item
$1,506.54 - $1,506.54
Specifications
Color White
Length (English) 32 in.
Length (Metric) 812mm
Material Polyurethane, CSM (Chlorosulfonated Polyethylene)
Resistant To High mechanical properties, puncture and tear resistance, highest performance with in situ leak detectors, high resistance to UV Rays/Ozone, natural aging, very resistant to reducing Acids, diluted Acids, Alkalis and Salts
View More Specs
Products 1
Catalog Number Mfr. No. Size Hand Port Size (English) Port Size (Metric) Price Quantity  
Catalog Number Mfr. No. Size Hand Port Size (English) Port Size (Metric) Price Quantity  
FSTP9773956 View Documents PIERCAN USA
7UY2032975A
9.75 Ambidextrous 7 in. 175mm Pair for $1,506.54
Specifications
Specifications
White
812mm
High mechanical properties, puncture and tear resistance, highest performance with in situ leak detectors, high resistance to UV Rays/Ozone, natural aging, very resistant to reducing Acids, diluted Acids, Alkalis and Salts
71°C
32 in.
Polyurethane, CSM (Chlorosulfonated Polyethylene)
160°F
20 mil