Please login to your online account to display your discounted pricing

Pictorial Periodic System of the Elements

Full-color photographs of the elements in their physical state help students retain faster

$58.75 - $58.75

Specifications

Classification Chemistry
For Use With (Application) Full color photographs of the elements in their physical state help students retain faster
View More Specs
Products
Catalog Number Mfr. No. Format Height (English) Packaging Quantity Width (English) Price Quantity    

S45562A

 
fisher science education
BINDERSIZE
Pack for $58.75
Description & Specifications

Specifications

Classification Chemistry
For Use With (Application) Full color photographs of the elements in their physical state help students retain faster

Full-color design of the periodic elements helps students learn all of the elements through visual representation.
Indicated for each element:
•    Physical state (whether it is solid, liquid, or gas)
•    Principal modifications
•    Metallic or nonmetallic character
•    Extreme reactivity to air and/or humidity
•    Characteristic use
•    Symbol, atomic number, atomic mass, group number, and period

Lecture-size chart is printed on highly durable plastic paper, in a large 2.5 × 1.5m format. It is shipped in four sections that can be easily assembled. The small poster measures 97 × 63cm. Also available in a convenient notebook-size version.