missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Picrotoxin, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 131210010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Picrotoxin | |
Authentic | |
Glass bottle | |
1g | |
− 32.00 | |
Solubility in water: 1g/5mL (100°C). Other solubilities: soluble in boiling alcohol (1gr/3mL),dilute acids,and alkalies; sparingly soluble in ether and ccl4,readily soluble in strong ammonia water and in aq.,solns of naoh.,1gr/13.5 mL in 95% ethan | |
CC(=C)C1C2C3C4(C(C1C(=O)O2)(CC5C4(O5)C(=O)O3)O)C.CC12C3C4C(C(C1(CC5C2(O5)C(=O)O3)O)C(=O)O4)C(C)(C)O | |
131846146 | |
98% |
124-87-8 | |
97.5% min. (Mixture of Picrotoxinin and Picrotin) (HPLC) | |
C30H34O13 | |
15,7527 | |
picrotoxin | |
VJKUPQSHOVKBCO-DMRKGALJSA-N | |
602.58 | |
602.58 |
Safety and Handling
GHS H Statement
Fatal if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 204-716-6
Recommended Storage : 4°C
RUO â Research Use Only