Learn More
Picolinic acid N-oxide, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 131180250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Picolinic acid N-oxide | |
824-40-8 | |
96% min. (HPLC) | |
C6H5NO3 | |
25g | |
FHYMLBVGNFVFBT-UHFFFAOYSA-N | |
1-oxidopyridin-1-ium-2-carboxylic acid | |
69986 | |
97% |
97% | |
Authentic | |
Glass bottle | |
MFCD00006198 | |
picolinic acid n-oxide, 2-carboxypyridin-1-ium-1-olate, pyridine-2-carboxylic acid 1-oxide, picolinic acid 1-oxide, 2-carboxypyridine n-oxide, 2-pyridinecarboxylic acid, 1-oxide, chembl73173, 2-pyridinecarboxylicacid, 1-oxide, picolinate n-oxide, 1-oxopicolinic acid | |
C1=CC=[N+](C(=C1)C(=O)O)[O-] | |
139.11 | |
139.11 |
Chemical Identifiers
824-40-8 | |
139.11 | |
FHYMLBVGNFVFBT-UHFFFAOYSA-N | |
69986 | |
C1=CC=[N+](C(=C1)C(=O)O)[O-] |
C6H5NO3 | |
MFCD00006198 | |
picolinic acid n-oxide, 2-carboxypyridin-1-ium-1-olate, pyridine-2-carboxylic acid 1-oxide, picolinic acid 1-oxide, 2-carboxypyridine n-oxide, 2-pyridinecarboxylic acid, 1-oxide, chembl73173, 2-pyridinecarboxylicacid, 1-oxide, picolinate n-oxide, 1-oxopicolinic acid | |
1-oxidopyridin-1-ium-2-carboxylic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
GHS Signal Word: Warning
EINECSNumber : 212-528-0
RUO â Research Use Only