Learn More
Picolinic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 131171000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Picolinic acid | |
98-98-6 | |
White | |
Authentic | |
Plastic bottle | |
MFCD00006293 | |
22, 33 | |
15, 7515 | |
Solubility in water: 887g/L (20°C). Other solubilities: soluble in ethanol (63g/L at 20°C),in glacial,acetic acid;,practically insoluble in ether,chloroform and,carbon disulfide | |
C1=CC=NC(=C1)C(=O)O | |
123.11 | |
CHEBI:28747 | |
99% |
99% | |
136°C to 139°C | |
3 to 4 (1% soln.) | |
99% | |
C6H5NO2 | |
100 g | |
05,538; 07,293 | |
picolinic acid, 2-pyridinecarboxylic acid, 2-picolinic acid, 2-carboxypyridine, alpha-picolinic acid, o-pyridinecarboxylic acid, pyridinecarboxylic acid, alpha-pyridinecarboxylic acid, picolinate, acide picolique | |
SIOXPEMLGUPBBT-UHFFFAOYSA-N | |
pyridine-2-carboxylic acid | |
1018 | |
123.11 | |
Crystalline Powder |
Chemical Identifiers
98-98-6 | |
123.11 | |
SIOXPEMLGUPBBT-UHFFFAOYSA-N | |
1018 | |
pyridine-2-carboxylic acid |
C6H5NO2 | |
MFCD00006293 | |
picolinic acid, 2-pyridinecarboxylic acid, 2-picolinic acid, 2-carboxypyridine, alpha-picolinic acid, o-pyridinecarboxylic acid, pyridinecarboxylic acid, alpha-pyridinecarboxylic acid, picolinate, acide picolique | |
CHEBI:28747 | |
C1=CC=NC(=C1)C(=O)O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if presen
GHS Signal Word: Warning
EINECSNumber : 202-719-7
RTECSNumber : TJ7344000
TSCA : TSCA
RUO – Research Use Only