missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Picloram, TCI America™

Manufacturer:  TCI America P18645G

 View more versions of this product

Catalog No. P18645G



Specifications

Specifications

Picloram
219°C
1918-02-1
MFCD00012101
picloram, 4-amino-3,5,6-trichloropicolinic acid, tordon, borolin, grazon, amdon, 2-pyridinecarboxylic acid, 4-amino-3,5,6-trichloro, k-pin, tordon 10k, tordon 22k
C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N
241.45
CHEBI:34922
Crystal-Powder at 20°C
White-Yellow
5g
C6H3Cl3N2O2
3077
NQQVFXUMIDALNH-UHFFFAOYSA-N
4-amino-3,5,6-trichloropyridine-2-carboxylic acid
15965
241.45
≥98.0% (HPLC)

Chemical Identifiers

1918-02-1
241.45
NQQVFXUMIDALNH-UHFFFAOYSA-N
15965
4-amino-3,5,6-trichloropyridine-2-carboxylic acid
C6H3Cl3N2O2
MFCD00012101
picloram, 4-amino-3,5,6-trichloropicolinic acid, tordon, borolin, grazon, amdon, 2-pyridinecarboxylic acid, 4-amino-3,5,6-trichloro, k-pin, tordon 10k, tordon 22k
CHEBI:34922
C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N
Safety and Handling

Safety and Handling

RTECSNumber : TJ7525000

TSCA : Yes

Documents