missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Picloram, TCI America™

Manufacturer:  TCI America P186425G

 View more versions of this product

Catalog No. P186425G



Specifications

Specifications

Picloram
219°C
1918-02-1
C6H3Cl3N2O2
3077
NQQVFXUMIDALNH-UHFFFAOYSA-N
4-amino-3,5,6-trichloropyridine-2-carboxylic acid
15965
241.45
White-Yellow
25g
≥98.0% (HPLC)
MFCD00012101
4-Amino-3,5,6-trichloropicolinic Acid, 4-Amino-3,5,6-trichloropyridine-2-carboxylic Acid
C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N
241.45
CHEBI:34922
Crystal-Powder at 20°C

Chemical Identifiers

1918-02-1
241.45
NQQVFXUMIDALNH-UHFFFAOYSA-N
15965
4-amino-3,5,6-trichloropyridine-2-carboxylic acid
C6H3Cl3N2O2
MFCD00012101
4-Amino-3,5,6-trichloropicolinic Acid, 4-Amino-3,5,6-trichloropyridine-2-carboxylic Acid
CHEBI:34922
C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N
Safety and Handling

Safety and Handling

RTECSNumber : TJ7525000

TSCA : Yes

Documents