Please login to your online account to display your discounted pricing

Piclamilast, Tocris™

Potent and selective PDE4 inhibitor

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material Piclamilast
Assay Percent Range >99%
Molecular Formula C18H18Cl2N2O3
Formula Weight 381.25
Synonym 3-(Cyclopentyloxy)-N-(3, 5-dichloro-4-pyridinyl)-4-methoxybenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

452510

 
r & d systems
RDS452510
10mg Each for $244.81

452550

 
r & d systems
RDS452550
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 381.25
Quantity 10mg
Synonym 3-(Cyclopentyloxy)-N-(3, 5-dichloro-4-pyridinyl)-4-methoxybenzamide
Chemical Name or Material Piclamilast
Assay Percent Range >99%
Molecular Formula C18H18Cl2N2O3
CAS 144035-83-6
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature