missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Piceid | |
226°C | |
C20H22O8 | |
5 g | |
HSTZMXCBWJGKHG-CUYWLFDKSA-N | |
(2S,3R,4S,5S,6R)-2-[3-hydroxy-5-[(E)-2-(4-hydroxyphenyl)ethenyl]phenoxy]-6-(hydroxymethyl)oxane-3,4,5-triol | |
5281718 | |
390.39 | |
Crystalline Powder |
27208-80-6 | |
White-Yellow | |
MFCD00210592 | |
Resveratrol 3-Glucoside, 4′,5-Dihydroxystilben-3-yl beta-D-Glucopyranoside, Polydatin | |
C1=CC(=CC=C1C=CC2=CC(=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O)O)O | |
390.388 | |
CHEBI:8198 | |
≥95.0% (HPLC) |
Chemical Identifiers
27208-80-6 | |
390.388 | |
HSTZMXCBWJGKHG-CUYWLFDKSA-N | |
CHEBI:8198 | |
C1=CC(=CC=C1C=CC2=CC(=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O)O)O |
C20H22O8 | |
MFCD00210592 | |
5281718 | |
(2S,3R,4S,5S,6R)-2-[3-hydroxy-5-[(E)-2-(4-hydroxyphenyl)ethenyl]phenoxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator