Please login to your online account to display your discounted pricing

Piceatannol, 96%, ACROS Organics™

$566.53 - $566.53

Chemical Identifiers

Chemical Name or Material Piceatannol
CAS 10083-24-6
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C14H12O4
Formula Weight 244.3
MDL Number MFCD00221715
Synonym 3,4,3',5'-Tetrahydroxy-trans-stilbene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC328450100

  SDS
thermo fisher scientific
328450100
10mg Each for $566.53
Description & Specifications

Specifications

Assay ≥95%
Physical Form Crystalline Powder
Formula Weight 244.3
Quantity 10mg
Color Yellow to golden or green
Chemical Name or Material Piceatannol
Assay Percent Range 96%
Molecular Formula C14H12O4
MDL Number MFCD00221715
CAS 10083-24-6
Synonym 3,4,3',5'-Tetrahydroxy-trans-stilbene
Infrared Spectrum Authentic