Please login to your online account to display your discounted pricing

Piceatannol, 96%, ACROS Organics™


Quantity 10mg

Manufacturer: thermo fisher scientific  328450100

 View more versions of this product

Catalog No. AC328450100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Crystalline Powder
Formula Weight 244.3
Quantity 10mg
Color Yellow to golden or green
Chemical Name or Material Piceatannol
Assay Percent Range 96%
Molecular Formula C14H12O4
MDL Number MFCD00221715
CAS 10083-24-6
Synonym 3,4,3',5'-Tetrahydroxy-trans-stilbene
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Piceatannol
CAS 10083-24-6
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C14H12O4
Formula Weight 244.3
MDL Number MFCD00221715
Synonym 3,4,3',5'-Tetrahydroxy-trans-stilbene