missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ PI4KB (PI4K beta) Recombinant Human Protein DFS Item

PI4KB and PI4KA are the two human type III PI4 kinases.

$1228.00 - $19320.00

Specifications

Accession Number NP_002642.1
Formulation 50mM Tris pH 7.5, 50% Glycerol, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT
Molecular Weight (g/mol) 120kDa
Storage Requirements Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Gene Alias PI4KB (PI4K beta)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV5277 DFS Item
View Documents Promotion Details
Invitrogen™
PV5277
20μg
Each for $1,228.00
 
PR9008A DFS Item
View Documents Promotion Details
Invitrogen™
PR9008A
100μg
Each for $2,665.00
 
PV5278 DFS Item
View Documents Promotion Details
Invitrogen™
PV5278
1mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

PI4KB and PI4KA share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol.

Specifications

Specifications

NP_002642.1
120kDa
PI4KB (PI4K beta)
Human
GST Tag
Frozen
50mM Tris pH 7.5, 50% Glycerol, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
PI3 Kinase Family
Recombinant
Baculovirus
SDS
missing translation for 'documents'

missing translation for 'documents'