Invitrogen™ PI4KB (PI4K beta) Recombinant Human Protein DFS Item
PI4KB and PI4KA are the two human type III PI4 kinases.
Manufacturer:  Invitrogen™ LSPV5278
 View more versions of this product
Catalog No. PV5278
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
PI4KB and PI4KA share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol.
Specifications
Specifications
NP_002642.1
Frozen
120kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
PI3 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris pH 7.5, 50% Glycerol, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT
1mg
RUO - research use only
PI4KB (PI4K beta)
Baculovirus
Human
GST Tag
Documents