Please login to your online account to display your discounted pricing

Phycoerythrin Mouse, Clone: eBioPE-DLF, eBioscience™

Mouse Monoclonal Antibody

$100.05 - $361.05

Specifications

Antigen Phycoerythrin
Clone eBioPE-DLF
Host Species Murine
Gene Alias Anti-PE
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013031

 
life technologies
LS14412085
500μg Each for $361.05

5013030

 
life technologies
LS14412081
50μg Each for $100.05
Description & Specifications

Specifications

Antigen Phycoerythrin
Clone eBioPE-DLF
Host Species Murine
Gene Alias Anti-PE
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The eBioPE-DLF monoclonal antibody reacts with phycoerythrin (PE), PE-Cy5 and PE-Cy7, with no crossreactivity to fluorescein (FITC) or allophycocyanin (APC). When using eBioPE-DLF as a second-step antibody, in combination with an appropriate third step, to bind a PE-conjugated primary antibody, there is no decrease in the fluorescent intensity of the directly conjugated PE antibody. Therefore no interference or quenching of the PE fluorochrome is observed. The eBioPE-DLF can be used for separation of cells labeled with PE-conjugated antibodies or staining. Please note that the optimal concentration for this antibody will vary depending on the density of the primary PE-conjugated antibody and the application tested.