missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phycoerythrin Mouse, Biotin, Clone: eBioPE-DLF, eBioscience™

Mouse Monoclonal Antibody

$64.68 - $143.88

Specifications

Antigen Phycoerythrin
Clone eBioPE-DLF
Host Species Murine
Gene Alias Anti-PE
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011847 Life Technologies
LS13412080
25μg Each for $64.68
501129375 Life Technologies
LS13412082
100μg Each for $143.88
Specifications

Description

The eBioPE-DLF monoclonal antibody reacts with phycoerythrin (PE), PE-Cy5 and PE-Cy7, with no crossreactivity to fluorescein (FITC) or allophycocyanin (APC). When using eBioPE-DLF as a second-step antibody, in combination with an appropriate third step, to bind a PE-conjugated primary antibody, there is no decrease in the fluorescent intensity of the directly conjugated PE antibody. Therefore no interference or quenching of the PE fluorochrome is observed. The eBioPE-DLF can be used for separation of cells labeled with PE-conjugated antibodies or staining. Please note that the optimal concentration for this antibody will vary depending on the density of the primary PE-conjugated antibody and the application tested.
Description & Specifications

Specifications

Phycoerythrin
Murine
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
eBioPE-DLF
Anti-PE
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer