Learn More
Thermo Scientific™ Phusion™ Plus PCR Master Mix
Thermo Scientific Phusion Plus PCR Master Mix is a convenient, ready-to-use 2X mixture of Phusion Plus DNA Polymerase, buffer, magnesium chloride, and dNTPs.
$307.00 - $4310.00
Specifications
GC-Rich PCR Performance | High |
---|---|
Polymerase | Phusion Plus DNA Polymerase |
Reaction Speed | 15 to 30 sec./kb |
Shipping Condition | Dry Ice |
For Use With (Application) | Hot-start PCR, High-fidelity PCR |
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
F631L
|
Thermo Scientific™
F631L |
500 Reactions |
Each for $1,200.00
|
|
|||||
F631S
|
Thermo Scientific™
F631S |
100 Reactions |
Each for $307.00
|
|
|||||
F631XL
|
Thermo Scientific™
F631XL |
2000 Reactions |
Each for $4,310.00
|
|
|||||
Description
Thermo Scientific Phusion Plus PCR Master Mix is a convenient, ready-to-use 2X mixture of Phusion Plus DNA Polymerase, buffer, magnesium chloride, and dNTPs. The master mix format simplifies reaction setup with minimal pipetting steps and retains all features of the stand-alone format.
• High fidelity—provides >100X greater sequence accuracy than Taq polymerase
• Convenient setup—enables a universal annealing temperature of 60°C in PCR
• Enhanced specificity—hot-start modification reduces non-specific amplification and primer degradation
• Benchtop stability—automation enabled due to 24-hour stability of assembled reactions
• High inhibitor tolerance—works with DNA of suboptimal purity
Phusion Plus DNA Polymerase is a hot-start, high-fidelity DNA polymerase that brings together protein fusion technology and universal primer annealing. It is a Pyrococcus-like proofreading DNA polymerase fused to a processivity-enhancing domain, enabling Phusion Plus PCR Master Mix to generate PCR sequences with high accuracy, sensitivity, and inhibitor tolerance. Its novel reaction buffer includes proprietary additives that stabilize primer/template duplexes during the annealing step at 60°C. This unique buffer formulation increases PCR specificity without the need to optimize annealing temperature for each primer pair, helping save time and avoid mistakes in PCR runs. Furthermore, with Phusion Plus PCR Master Mix and its universal annealing feature, different PCR reactions can be cycled together using the same protocol.
Phusion Plus PCR Master Mix is inactive at ambient temperatures due to its hot-start mechanism mediated by Affibody molecules, allowing setup and storage of assembled PCR reactions at room temperature. Enzyme activity is fully restored after the initial denaturation step.
Applications
• High-fidelity PCR
• Cloning
• Template generation for sequencing
• GC-rich PCR
• Long-range PCR (up to 20 kb)
• Mutagenesis
• High-throughput PCR
Specifications
High | |
15 to 30 sec./kb | |
Hot-start PCR, High-fidelity PCR | |
>100X | |
Blunt | |
20 kb or less |
Phusion Plus DNA Polymerase | |
Dry Ice | |
2X | |
Built-In Hot Start | |
Ready to use master mix |
For Research Use Only. Not for use in diagnostic procedures.