Please login to your online account to display your discounted pricing

Phthaloyl chloride, 90%, Acros Organics


Quantity 100mL
Packaging AcroSeal™ Glass bottle

Manufacturer: thermo fisher scientific  AC417971000

 View more versions of this product

Catalog No. AC417971000

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Phthaloyl chloride
Quantity 100mL
CAS 88-95-9
Packaging AcroSeal™ Glass bottle
Assay Percent Range 90%
Molecular Formula C8H4Cl2O2
Formula Weight 203.02
Synonym 1, 2-Benzenedicarbonyl dichloride

Specifications

Chemical Name or Material Phthaloyl chloride
Assay Percent Range 90%
Molecular Formula C8H4Cl2O2
Formula Weight 203.02
Synonym 1, 2-Benzenedicarbonyl dichloride