Please login to your online account to display your discounted pricing

Phthaloyl chloride, 90%, Acros Organics

$59.36 - $59.36

Chemical Identifiers

Chemical Name or Material Phthaloyl chloride
Assay Percent Range 90%
Molecular Formula C8H4Cl2O2
Formula Weight 203.02
Synonym 1, 2-Benzenedicarbonyl dichloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC417971000

  SDS
thermo fisher scientific
AC417971000
AcroSeal™ Glass bottle 100mL Each for $59.36

AC417978000

  SDS
thermo fisher scientific
AC417978000
AcroSeal™ Glass bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.02
Quantity 100mL
Assay Percent Range 90%
Molecular Formula C8H4Cl2O2
Chemical Name or Material Phthaloyl chloride
Packaging AcroSeal™ Glass bottle
Synonym 1, 2-Benzenedicarbonyl dichloride
CAS 88-95-9