Please login to your online account to display your discounted pricing

Phthaloyl chloride, 90%, Acros Organics


Quantity 800mL
Packaging AcroSeal™ Glass bottle

Manufacturer: thermo fisher scientific  AC417978000

 View more versions of this product

Catalog No. AC417978000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Phthaloyl chloride
Quantity 800mL
CAS 88-95-9
Packaging AcroSeal™ Glass bottle
Assay Percent Range 90%
Molecular Formula C8H4Cl2O2
Formula Weight 203.02
Synonym 1, 2-Benzenedicarbonyl dichloride

Specifications

Chemical Name or Material Phthaloyl chloride
Assay Percent Range 90%
Molecular Formula C8H4Cl2O2
Formula Weight 203.02
Synonym 1, 2-Benzenedicarbonyl dichloride