missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phthalonitrile, 98%

Manufacturer:  Alfa Aesar™ A1507122*

 View more versions of this product

Catalog No. AAA1507122



Description

Description

Applications
Phthalonitrile is used as an intermediate for chemical synthesis in the production of building blocks for colorants and coatings, life science and agricultural chemicals. And also used as a precursor to phthalocyanine and other pigments, fluorescent brighteners, and photographic sensitizers.

Solubility
Soluble in alcohol and ether.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Phthalonitrile
139°C to 141°C
91-15-6
MFCD00001771
775028
XQZYPMVTSDWCCE-UHFFFAOYSA-N
benzene-1,2-dicarbonitrile
7042
98%
162°C (323°F)
100g
C8H4N2
UN3439
phthalonitrile, 1,2-dicyanobenzene, o-phthalodinitrile, phthalodinitrile, 1,2-benzenedicarbonitrile, o-dicyanobenzene, 1,2-benzodinitrile, o-pdn, o-cyanobenzonitrile, o-benzenedicarbonitrile
C1=CC=C(C(=C1)C#N)C#N
128.134
128.13

Chemical Identifiers

91-15-6
128.134
XQZYPMVTSDWCCE-UHFFFAOYSA-N
7042
C1=CC=C(C(=C1)C#N)C#N
C8H4N2
MFCD00001771
phthalonitrile, 1,2-dicyanobenzene, o-phthalodinitrile, phthalodinitrile, 1,2-benzenedicarbonitrile, o-dicyanobenzene, 1,2-benzodinitrile, o-pdn, o-cyanobenzonitrile, o-benzenedicarbonitrile
benzene-1,2-dicarbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H300-H311
Fatal if swallowed.
Toxic in contact with skin.

GHS P Statement
P280-P301+P310-P361-P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

RTECSNumber : TI8575000

TSCA : TSCA

Documents