missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phthalocyanine

Manufacturer:  Alfa Aesar™ 3207306

 View more versions of this product

Catalog No. AA3207306



Specifications

Specifications

Phthalocyanine
5g
C32H18N8
IEQIEDJGQAUEQZ-UHFFFAOYSA-N
514.552
CHEBI:34921
Crystalline
Odorless
574-93-6
MFCD00005085
C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C(N4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2N3
5282330
514.52

Chemical Identifiers

574-93-6
514.552
IEQIEDJGQAUEQZ-UHFFFAOYSA-N
CHEBI:34921
C32H18N8
MFCD00005085
5282330
C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C(N4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2N3
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents