missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phthalide, 98+%

Manufacturer:  Alfa Aesar™ A155160E

 View more versions of this product

Catalog No. AAA155160E



Specifications

Specifications

Phthalide
289°C to 291°C
2500g
C8H6O2
114632
WNZQDUSMALZDQF-UHFFFAOYSA-N
3H-2-benzofuran-1-one
6885
134.13
152°C (305°F)
72°C to 75°C
87-41-2
MFCD00005906
phthalide, isobenzofuran-1 3h-one, 1 3h-isobenzofuranone, 1-phthalanone, phthalolactone, 2-benzofuran-1 3h-one, isobenzofuranone, phthalanone, 3h-isobenzofuran-1-one, 2-hydroxymethylbenzoic acid, gamma-lactone
C1C2=CC=CC=C2C(=O)O1
134.134
CHEBI:38085
≥98%

Chemical Identifiers

87-41-2
134.134
WNZQDUSMALZDQF-UHFFFAOYSA-N
6885
3H-2-benzofuran-1-one
C8H6O2
MFCD00005906
phthalide, isobenzofuran-1 3h-one, 1 3h-isobenzofuranone, 1-phthalanone, phthalolactone, 2-benzofuran-1 3h-one, isobenzofuranone, phthalanone, 3h-isobenzofuran-1-one, 2-hydroxymethylbenzoic acid, gamma-lactone
CHEBI:38085
C1C2=CC=CC=C2C(=O)O1
Safety and Handling

Safety and Handling

RTECSNumber : TI3520000

TSCA : TSCA

Documents