Please login to your online account to display your discounted pricing

Alfa Aesar™ Phthalide-3-acetic acid, 98+%

$134.24 - $134.24

Chemical Identifiers

Chemical Name or Material Phthalide-3-acetic acid
CAS 4743-58-2
Molecular Formula C10H8O4
Formula Weight 192.17
MDL Number MFCD00051729
Beilstein 156005
Synonym 1(3H)-Isobenzofuranone-3-acetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0598414

  SDS
alfa aesar™
L0598414
25g Each for $134.24
Description & Specifications

Specifications

Formula Weight 192.17
Chemical Name or Material Phthalide-3-acetic acid
MDL Number MFCD00051729
Health Hazard 1 Warning
Molecular Formula C10H8O4
Assay ≥98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 156005
Quantity 25g
Melting Point 148° to 153°C
CAS 4743-58-2
Synonym 1(3H)-Isobenzofuranone-3-acetic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.