Please login to your online account to display your discounted pricing

Phthalic Anhydride, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: avantor performance  027205

 View more versions of this product

Catalog No. 02003867

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 148.12
CAS 85-44-9
Chemical Name or Material Phthalic Anhydride
Molecular Formula C8H4O3
Synonym 1,1-Benzene Dicarboxilic Acid
Packaging Poly Bottle
Flash Point 152.00°C
Quantity 2.5kg
Specific Gravity 1L = 1.53kg

Specifications

Chemical Name or Material Phthalic Anhydride
CAS 85-44-9
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula C8H4O3
Formula Weight 148.12
Synonym 1,1-Benzene Dicarboxilic Acid