Please login to your online account to display your discounted pricing

Phthalic Anhydride, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$87.05 - $173.41

Chemical Identifiers

Chemical Name or Material Phthalic Anhydride
CAS 85-44-9
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula C8H4O3
Formula Weight 148.12
Synonym 1,1-Benzene Dicarboxilic Acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003868

 
avantor performance
027201
Poly Bottle 500g Each for $87.05

02003867

 
avantor performance
027205
Poly Bottle 2.5kg Each for $173.41
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 148.12
Chemical Name or Material Phthalic Anhydride
CAS 85-44-9
Packaging Poly Bottle
Molecular Formula C8H4O3
Synonym 1,1-Benzene Dicarboxilic Acid
Flash Point 152.00°C
Specific Gravity 1L = 1.53kg
Quantity 500g