Please login to your online account to display your discounted pricing

Phthalic Anhydride, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  027201

 View more versions of this product

Catalog No. 02003868

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 148.12
Purity Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Phthalic Anhydride
CAS 85-44-9
Synonym 1,1-Benzene Dicarboxilic Acid
Quantity 500g
Packaging Poly Bottle
Molecular Formula C8H4O3
Flash Point 152.00°C
Specific Gravity 1L = 1.53kg

Specifications

Chemical Name or Material Phthalic Anhydride
CAS 85-44-9
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula C8H4O3
Formula Weight 148.12
Synonym 1,1-Benzene Dicarboxilic Acid