Learn More
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 154962500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phthalic anhydride | |
129.0°C to 132.0°C | |
152°C | |
Plastic bottle | |
MFCD00005918 | |
17,469 | |
15,7484 | |
Solubility in water: 6g/L (20°C). Other solubilities: soluble in alcohol and carbon disulfide, slightly soluble in ether, 7.5% in benzene (25°C), forms phthalic acid in water | |
O=C1OC(=O)C2=CC=CC=C12 | |
148.12 | |
6.4 mPa.s (197°C) | |
148.12 |
85-44-9 | |
284.0°C | |
99% | |
C8H4O3 | |
250 g | |
01,882; 17,148 | |
phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen | |
LGRFSURHDFAFJT-UHFFFAOYSA-N | |
2-benzofuran-1,3-dione | |
6811 | |
CHEBI:36605 | |
99% |
Chemical Identifiers
85-44-9 | |
148.12 | |
LGRFSURHDFAFJT-UHFFFAOYSA-N | |
6811 | |
O=C1OC(=O)C2=CC=CC=C12 |
C8H4O3 | |
MFCD00005918 | |
phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen | |
CHEBI:36605 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allerg
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat
GHS Signal Word: Danger
EINECSNumber : 201-607-5
RUO – Research Use Only