Please login to your online account to display your discounted pricing

Phthalhydrazide 99%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  131061000

 View more versions of this product

Catalog No. AC131061000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder and Chunks
Formula Weight 162.15
Quantity 100g
Color White
Beilstein 24, 371
EINECS Number 215-893-4
Assay 98.5 to 101.5% (on dried substance)
RTECS Number TH8890080
Infrared Spectrum Authentic
Chemical Name or Material Phthalhydrazide
Packaging Plastic bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8H6N2O2
MDL Number MFCD00006888
CAS 1445-69-8
Synonym 2, 3-Dihydro-1, 4-phthalazinedione
Loss on Drying 1% max.

Specifications

Chemical Name or Material Phthalhydrazide
Name Note 99%
CAS 1445-69-8
Assay 98.5 to 101.5% (on dried substance)
Assay Percent Range 99%
Molecular Formula C8H6N2O2
Formula Weight 162.15
MDL Number MFCD00006888
Synonym 2, 3-Dihydro-1, 4-phthalazinedione