Please login to your online account to display your discounted pricing

Alfa Aesar™ Phthalhydrazide, 98%

$28.78 - $189.34

Chemical Identifiers

Chemical Name or Material Phthalhydrazide
CAS 1445-69-8
Molecular Formula C8H6N2O2
Formula Weight 162.15
MDL Number MFCD00006888
Beilstein 135926
Synonym 2,3-Dihydro-1,4-phthalazinedione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1461509

  SDS
alfa aesar™
A1461509
10g Each for $28.78

AAA1461530

  SDS
alfa aesar™
A1461530
250g Each for $189.34
Description & Specifications

Specifications

Formula Weight 162.15
Chemical Name or Material Phthalhydrazide
Assay 98%
Molecular Formula C8H6N2O2
MDL Number MFCD00006888
CAS 1445-69-8
Beilstein 135926
Synonym 2,3-Dihydro-1,4-phthalazinedione
RTECS Number TH8890080
Melting Point 342° to 345°C
Quantity 10g

Applications
Phthalhydrazide is a reagent used in the synthesis of various pyrazolophthalazine derivatives is and used in medicine double hydrazine phthalocyanine work intermediates.

Solubility
Soluble in acetone and acetic acid.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.