missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phthalazine 98%, ACROS Organics™

Chemical Identifiers

CAS 253-52-1
Assay Percent Range 98%
Molecular Formula C8H6N2
Formula Weight 130.15
MDL Number MFCD00006908
Synonym 2, 3-Benzodiazine
Chemical Name or Material Phthalazine, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC131050100 SDS Fisher Scientific Acros
131050100
Glass bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

253-52-1
C8H6N2
MFCD00006908
Phthalazine, 98%
98%
130.15
2, 3-Benzodiazine
≥97.5%

Specifications

130.15
10g
88° to 92°C
15, 7482
Glass bottle
98%
C8H6N2
Phthalazine, 98%
Crystalline Powder
Slightly brown to beige
23, 174
Authentic
2, 3-Benzodiazine
MFCD00006908
253-52-1
≥97.5%

Safety and Handling

EINECSNumber : 205-963-2

TSCA : TSCA