Please login to your online account to display your discounted pricing

Phthalates in PVC QC Standards, Spex CertiPrep

Matrix: Poly(vinyl) Chloride

$351.77 - $351.77

Chemical Identifiers

Name Note Designed to meet the regulations required for testing of PVC toys and other consumer products
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CRMPVC001

 
spex certiprep llc
SPXCRMPVC001
2mL amber prescored glass ampule 1.5g Each for $351.77
Description & Specifications

Specifications

Quantity 1.5g
DOT Information Nonhazardous
Chemical Name or Material Phthalates in Polyvinyl Chloride
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Packaging 2mL amber prescored glass ampule
Recommended Storage Ambient
Methods For Consumer Product Safety Commission Method CPSC-CH-C1001-09.1
Solvent or Matrix Polyvinyl Chloride
Di n butyl phthalate (DBP) 3000μg/g
Diethyl phthalate (DEP) 3000μg/g
Dimethyl phthalate (DMP) 3000μg/g
Di n octyl phthalate (DOP) 3000μg/g
Butylbenzyl Phthalate (BBP) 3000μg/g
Name Note Designed to meet the regulations required for testing of PVC toys and other consumer products
Diisononyl phthalate (DINP) 30000μg/g
bis(2 Ethylhexyl) phthalate (DEHP) 3000μg/g
Diisodecyl phthalate (DIDP) 30000μg/g

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Ambient