Please login to your online account to display your discounted pricing

Alfa Aesar™ Phthalamidoxime, 97%

Chemical Identifiers

Chemical Name or Material Phthalamidoxime
CAS 885960-32-7
Formula Weight 194.19
Synonym 1,2-Diamidoximobenzene; Benzene-1,2-dicarboxamidoxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6634203

 
alfa aesar™
H6634203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66342MD

 
alfa aesar™
H66342MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.19
Chemical Name or Material Phthalamidoxime
Assay 97%
Physical Form Solid
Synonym 1,2-Diamidoximobenzene; Benzene-1,2-dicarboxamidoxime
Color Cream
Quantity 1g
CAS 885960-32-7