missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Oakton™ Testr™ Meter Accessories
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Supplier: Cole-Parmer™ 3563447
Requires : WD-35634-45
Description
Handy replacement and accessory itemsWarranty
1 Year
Specifications
| ±2 mV ±1 LSD | |
| Multipin | |
| 3.5 x 1.1 in. | |
| 3.5 x 1.1 x 1.1 in. | |
| Water quality testing, buffer and reagent production, electrochemistry | |
| 3.5 in. | |
| No cable | |
| ORP sensor | |
| ±1000 mV | |
| 1 mV | |
| Manual | |
| 41°F to 113°F | |
| 0°C to 60°C | |
| 0.1 lb. | |
| 0.05 kg | |
| 2.8 cm | |
| Oakton ORPTestr 50 |
| Gray | |
| Replacement ORP sensor | |
| Replacement ORP Sensor | |
| Four AAA batteries, installed in meter | |
| ORP Sensor | |
| 8.89 cm | |
| Valox™ | |
| ORP | |
| â1000 to 1000 mV | |
| 0.1°F/°C | |
| 32°F to 140°F | |
| 60°C | |
| 5°C to 45°C | |
| 6 Months | |
| 1.1 in. | |
| None |